siRNA Details
| Gene Symbol: Tprgl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026388.2 | NP_080664.1 | 4 | 684 | ||
| GenBank mRNA | AK004324.1 | - | 3 | 297 | ||
| AK004777.1 | - | 4 | 667 | |||
| AK155413.1 | - | 4 | 679 | |||
| AK176961.1 | - | 3 | 232 | |||
| AK178047.1 | - | 1 | 15 | |||
| AK178710.1 | - | 2 | 78 | |||
| AK189153.1 | - | 2 | 78 | |||
| AK195680.1 | - | 2 | 134 | |||
| AK196897.1 | - | 3 | 285 | |||
| AK206032.1 | - | 2 | 134 | |||
| AK207196.1 | - | 2 | 134 | |||
| AK209255.1 | - | 2 | 134 | |||
| AK210976.1 | - | 2 | 134 | |||
| AK211127.1 | - | 2 | 134 | |||
| AK213307.1 | - | 2 | 134 | |||
| AK213716.1 | - | 3 | 233 | |||
| AK214091.1 | - | 2 | 78 | |||
| AK218094.1 | - | 2 | 134 | |||
| AK220054.1 | - | 2 | 134 | |||
| BC019544.1 | - | 4 | 648 | 4211201 | ||

Tprgl
Genomic Map