siRNA Details
| Gene Symbol: ITGA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000210.3 | NP_000201.2 | 1 | 332 | ||
| NM_001079818.2 | NP_001073286.1 | 1 | 332 | |||
| XM_006712510.1 | XP_006712573.1 | 1 | 301 | |||
| XM_006712511.1 | XP_006712574.1 | 1 | 301 | |||
| GenBank mRNA | BC007002.1 | - | 1 | 298 | ||
| BC136455.1 | - | 1 | 206 | 9020444 | ||
| BC136456.1 | - | 1 | 206 | 9020445 | ||
| DA760250.1 | - | 1 | 301 | |||
| DB457895.1 | - | 1 | 332 | |||
| X53586.1 | - | 1 | 244 | |||
| X59512.1 | - | 1 | 80 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008397, NM_001277970, BC049132, X69902, BC058095, and others.

ITGA6
Genomic Map