siRNA Details
| Gene Symbol: ITGB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000212.2 | NP_000203.2 | 3 | 229 | ||
| GenBank mRNA | AY826979.1 | - | 3 | 218 | ||
| BC127666.1 | - | 3 | 229 | 40128462 | ||
| BC127667.1 | - | 3 | 233 | 40128464 | ||
| J02703.1 | - | 3 | 229 | |||
| KC120777.1 | - | 3 | 229 | |||
| KC120778.1 | - | 3 | 229 | |||
| M20311.1 | - | 3 | 225 | |||
| M35999.1 | - | 3 | 227 | |||
| U95204.1 | - | 3 | 209 | |||

ITGB3
Genomic Map