siRNA Details
| Gene Symbol: Tmem100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026433.2 | NP_080709.1 | 3 | 950 | ||
| GenBank mRNA | AK003574.2 | - | 3 | 577 | ||
| AK007878.1 | - | 3 | 951 | |||
| BC034841.1 | - | 2 | 565 | 3326079 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem100| Gene Symbol: Tmem100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026433.2 | NP_080709.1 | 3 | 950 | ||
| GenBank mRNA | AK003574.2 | - | 3 | 577 | ||
| AK007878.1 | - | 3 | 951 | |||
| BC034841.1 | - | 2 | 565 | 3326079 | ||