siRNA Details
| Gene Symbol: Ufm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026435.5 | NP_080711.1 | 6 | 612 | ||
| GenBank mRNA | AK007433.1 | - | 1 | 259 | ||
| AK007788.1 | - | 6 | 576 | |||
| AK008280.1 | - | 6 | 574 | |||
| AK089948.1 | - | 6 | 578 | |||
| AK148146.1 | - | 6 | 582 | |||
| AK166332.1 | - | 6 | 606 | |||
| AK167301.1 | - | 6 | 581 | |||
| AK167321.1 | - | 6 | 576 | |||
| BC061065.1 | - | 6 | 611 | 6594260 | ||

Ufm1
Genomic Map