siRNA Details
| Gene Symbol: Rgs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026380.3 | NP_080656.2 | 7 | 5269 | ||
| GenBank mRNA | AK018215.1 | - | 1 | 1062 | ||
| AK018337.1 | - | 1 | 1245 | |||
| BC056441.1 | - | 1 | 2275 | |||
| BC057559.1 | - | 1 | 1701 | |||
| BC065056.1 | - | 7 | 5202 | 5705033 | ||
| BC094577.1 | - | 6 | 4885 | 6852696 | ||

Rgs8
Genomic Map