siRNA Details
| Gene Symbol: Snrnp48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026382.2 | NP_080658.2 | 2, 3 | 306 | ||
| GenBank mRNA | AK007431.1 | - | 2, 3 | 295 | ||
| AK012916.1 | - | 2, 3 | 301 | |||
| AK018315.1 | - | 2, 3 | 301 | |||
| AK036713.1 | - | 2, 3 | 306 | |||
| AK172155.1 | - | 2, 3 | 295 | |||
| BC021443.1 | - | 2, 3 | 289 | 5038605 | ||
| BC037073.1 | - | 2, 3 | 296 | |||
| BY263553.1 | - | 2, 3 | 309 | |||

Snrnp48
Genomic Map