siRNA Details
| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290761.1 | NP_001277690.1 | 5 | 1162 | ||
| NM_001290762.1 | NP_001277691.1 | 5 | 1239 | |||
| NM_026393.2 | NP_080669.1 | 6 | 1192 | |||
| GenBank mRNA | AA166262.1 | - | 1 | 228 | ||
| AK012340.1 | - | 5 | 995 | |||
| AK146521.1 | - | 918 | ||||
| BC030039.1 | - | 6 | 895 | 1313824 | ||

Nmral1
Genomic Map