siRNA Details
Gene Symbol: Dnajb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190804.1 | NP_001177733.1 | 4, 5 | 647 | ||
NM_001190805.1 | NP_001177734.1 | 4, 5 | 647 | |||
NM_026400.5 | NP_080676.3 | 4, 5 | 647 | |||
GenBank mRNA | AK050500.1 | - | 4, 5 | 606 | ||
AK075785.1 | - | 4, 5 | 629 | |||
AK075953.1 | - | 4, 5 | 609 | |||
AK148511.1 | - | 4, 5 | 627 | |||
AK166730.1 | - | 4, 5 | 605 | |||
AY054981.1 | - | 4, 5 | 523 | |||
BC003999.2 | - | 4, 5 | 578 | 3492402 | ||
BC018282.1 | - | 4, 5 | 574 | 4207201 | ||
BC040747.1 | - | 4, 5 | 578 | 3492402 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
