siRNA Details
| Gene Symbol: Mrpl57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026401.2 | NP_080677.1 | 2 | 617 | ||
| GenBank mRNA | AK011558.1 | - | 2 | 618 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl57| Gene Symbol: Mrpl57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026401.2 | NP_080677.1 | 2 | 617 | ||
| GenBank mRNA | AK011558.1 | - | 2 | 618 | ||