siRNA Details
| Gene Symbol: 1700021F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026411.1 | NP_080687.1 | 4 | 705 | ||
| GenBank mRNA | AK006199.1 | - | 4 | 705 | ||
| AK007370.1 | - | 4 | 709 | |||
| AK008795.1 | - | 4 | 669 | |||
| AK163304.1 | - | 4 | 673 | |||
| BC013506.1 | - | 4 | 679 | 4224442 | ||

1700021F05Rik
Genomic Map