siRNA Details
| Gene Symbol: Enoph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026421.3 | NP_080697.2 | 6 | 1102 | ||
| GenBank mRNA | AK009954.1 | - | 6 | 1085 | ||
| AK079062.1 | - | 6 | 1096 | |||
| AK082747.1 | - | 6 | 1077 | |||
| AK152021.1 | - | 6 | 1076 | |||
| AK159356.1 | - | 6 | 1075 | |||
| AK167575.1 | - | 6 | 1113 | |||
| AK168696.1 | - | 6 | 971 | |||
| BC021429.1 | - | 6 | 1029 | 5030617 | ||

Enoph1
Genomic Map