siRNA Details
| Gene Symbol: Coq10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039710.1 | NP_001034799.1 | 3 | 422 | ||
| NM_026424.3 | NP_080700.3 | 2 | 272 | |||
| GenBank mRNA | AK002294.1 | - | 3 | 323 | ||
| AK006551.1 | - | 2 | 273 | |||
| AK012117.1 | - | 3 | 390 | |||
| AK015454.1 | - | 3 | 423 | |||
| AK136258.1 | - | 3 | 322 | |||
| AK146260.1 | - | 3 | 316 | |||
| AK168293.1 | - | 3 | 316 | |||
| AK168837.1 | - | 3 | 320 | |||
| BC062129.1 | - | 3 | 318 | 6506502 | ||

Coq10b
Genomic Map