siRNA Details
| Gene Symbol: Ufm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026435.5 | NP_080711.1 | 6 | 695 | ||
| GenBank mRNA | AK007433.1 | - | 1 | 342 | ||
| AK007788.1 | - | 6 | 659 | |||
| AK008280.1 | - | 6 | 657 | |||
| AK089948.1 | - | 6 | 661 | |||
| AK148146.1 | - | 6 | 664 | |||
| AK166332.1 | - | 6 | 689 | |||
| AK167301.1 | - | 6 | 664 | |||
| AK167321.1 | - | 6 | 659 | |||
| BC061065.1 | - | 6 | 694 | 6594260 | ||

Ufm1
Genomic Map