siRNA Details
| Gene Symbol: Cmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026442.3 | NP_080718.1 | 4 | 423 | ||
| GenBank mRNA | AK002667.1 | - | 4 | 320 | ||
| AK020626.1 | - | 4 | 307 | |||
| AK168728.1 | - | 4 | 370 | |||
| BC048547.1 | - | 4 | 404 | |||
| BC056653.1 | - | 4 | 297 | 5068125 | ||

Cmc1
Genomic Map