siRNA Details
| Gene Symbol: Mtfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026443.4 | NP_080719.2 | 4 | 1177 | ||
| GenBank mRNA | AK006148.1 | - | 4 | 1098 | ||
| AK010971.1 | - | 4 | 1134 | |||
| AK012058.1 | - | 4 | 1144 | |||
| AK078381.1 | - | 4 | 1178 | |||
| AK167990.1 | - | 4 | 1045 | |||
| AK216258.1 | - | 1 | 147 | |||
| BC012242.1 | - | 1 | 591 | |||
| BC019205.1 | - | 4 | 1187 | |||
| BC058737.1 | - | 4 | 1127 | 5400435 | ||

Mtfp1
Genomic Map