siRNA Details
| Gene Symbol: Ppm1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026447.4 | NP_080723.3 | 6 | 936 | ||
| NM_198931.3 | NP_945149.2 | 6 | 940 | |||
| GenBank mRNA | AB474373.1 | - | 6 | 975 | ||
| AK013149.1 | - | 6 | 869 | |||
| AK017245.1 | - | 3 | 273 | |||
| AK046387.1 | - | 835 | ||||
| AK087999.1 | - | 6 | 974 | |||
| AK216174.1 | - | 1 | 38 | |||
| AY332616.1 | - | 6 | 943 | |||
| BC058248.1 | - | 6 | 862 | |||

Ppm1m
Genomic Map