siRNA Details
| Gene Symbol: Fam32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026455.4 | NP_080731.1 | 3 | 305 | ||
| GenBank mRNA | AB041658.1 | - | 3 | 303 | ||
| AK007524.1 | - | 3 | 303 | |||
| AK008285.1 | - | 3 | 284 | |||
| AK008336.1 | - | 3 | 291 | |||
| AK011922.1 | - | 3 | 291 | |||
| AK075976.1 | - | 3 | 306 | |||
| AK082792.1 | - | 3 | 276 | |||
| AK150416.1 | - | 3 | 276 | |||
| BC006058.1 | - | 3 | 257 | 3591395 | ||
| BC085487.1 | - | 2 | 2116 | |||

Fam32a
Genomic Map