siRNA Details
| Gene Symbol: Ccdc70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026459.3 | NP_080735.1 | 2 | 489 | ||
| GenBank mRNA | AK006920.1 | - | 1 | 382 | ||
| AK007190.1 | - | 2 | 421 | |||
| BC049553.1 | - | 2 | 489 | 6773824 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc70| Gene Symbol: Ccdc70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026459.3 | NP_080735.1 | 2 | 489 | ||
| GenBank mRNA | AK006920.1 | - | 1 | 382 | ||
| AK007190.1 | - | 2 | 421 | |||
| BC049553.1 | - | 2 | 489 | 6773824 | ||