siRNA Details
| Gene Symbol: Tppp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026481.3 | NP_080757.1 | 2, 3 | 346 | ||
| GenBank mRNA | AK009771.1 | - | 2, 3 | 310 | ||
| AK012437.1 | - | 2, 3 | 314 | |||
| AK155018.1 | - | 2, 3 | 317 | |||
| AK160507.1 | - | 2, 3 | 317 | |||
| BC010788.1 | - | 2, 3 | 303 | 4216285 | ||
| BC116622.1 | - | 1, 2 | 138 | |||
| BF469439.1 | - | 2, 3 | 346 | |||

Tppp3
Genomic Map