siRNA Details
| Gene Symbol: Mphosph10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026483.2 | NP_080759.2 | 6, 7 | 1348 | ||
| GenBank mRNA | AK008785.1 | - | 3, 4 | 278 | ||
| AK013337.1 | - | 6, 7 | 1336 | |||
| BC018177.1 | - | 6, 7 | 1342 | |||
| BC049270.1 | - | 6, 7 | 1324 | 3988645 | ||

Mphosph10
Genomic Map