siRNA Details
| Gene Symbol: Grhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026496.4 | NP_080772.2 | 2 | 461 | ||
| GenBank mRNA | AF411213.1 | - | 2 | 194 | ||
| AK002633.1 | - | 2 | 462 | |||
| BC055035.1 | - | 2 | 250 | 6306106 | ||
| BY703140.1 | - | 2 | 462 | |||

Grhl2
Genomic Map