siRNA Details
| Gene Symbol: Cdc42ep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026514.2 | NP_080790.1 | 2 | 691 | ||
| GenBank mRNA | AK012309.1 | - | 2 | 656 | ||
| AK014281.1 | - | 2 | 693 | |||
| AK019447.1 | - | 2 | 693 | |||
| AK076782.1 | - | 2 | 490 | |||
| AK150214.1 | - | 2 | 658 | |||
| BC021409.1 | - | 1 | 345 | 5007192 | ||
| BC034714.1 | - | 2 | 644 | 3595315 | ||
| BC092297.1 | - | 2 | 640 | 5063754 | ||

Cdc42ep3
Genomic Map