siRNA Details
| Gene Symbol: MBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000242.2 | NP_000233.1 | 2 | 338 | ||
| XM_006717861.3 | XP_006717924.1 | 3 | 406 | |||
| XM_011539816.2 | XP_011538118.1 | 3 | 390 | |||
| GenBank mRNA | AF360991.1 | - | 2 | 289 | ||
| AY826184.1 | - | 2 | 273 | |||
| BC069338.1 | - | 2 | 323 | 7262231 | ||
| BC096179.3 | - | 2 | 304 | 40003735 | ||
| BC096180.3 | - | 2 | 304 | 40003741 | ||
| BC096181.3 | - | 2 | 304 | 40003742 | ||
| BC096182.3 | - | 2 | 304 | 40003743 | ||
| X15422.1 | - | 2 | 338 | |||

MBL2
Genomic Map