siRNA Details
| Gene Symbol: Rps13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026533.3 | NP_080809.1 | 4 | 277 | ||
| GenBank mRNA | AK012479.1 | - | 4 | 222 | ||
| AK018703.1 | - | 4 | 221 | |||
| AK076060.1 | - | 4 | 221 | |||
| AK088237.1 | - | 3 | 854 | |||
| AK151043.1 | - | 4 | 222 | |||
| AK151671.1 | - | 4 | 223 | |||
| AK161798.1 | - | 3 | 820 | |||
| AK166691.1 | - | 4 | 222 | |||
| AK177445.1 | - | 220 | ||||
| AV498110.1 | - | 3 | 184 | |||
| BC011192.1 | - | 4 | 220 | 4016679 | ||
| BC085269.1 | - | 2 | 926 | |||
| BC090397.1 | - | 4 | 204 | 5150054 | ||
| BC106137.1 | - | 4 | 205 | 4990874 | ||
| BC125602.1 | - | 4 | 221 | 40130117 | ||
| BC125604.1 | - | 4 | 221 | 40130119 | ||

Rps13
Genomic Map