siRNA Details
| Gene Symbol: 3010026O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026543.3 | NP_080819.2 | 1, 2 | 109 | ||
| GenBank mRNA | AK003636.1 | - | 1, 2 | 84 | ||
| AK019392.1 | - | 1, 2 | 110 | |||
| BC028428.1 | - | 1, 2 | 94 | 1315017 | ||

3010026O09Rik
Genomic Map