siRNA Details
| Gene Symbol: Yeats4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026570.4 | NP_080846.1 | 2 | 301 | ||
| GenBank mRNA | AK010522.1 | - | 2 | 302 | ||
| AK012096.1 | - | 2 | 268 | |||
| AK081867.1 | - | 2 | 284 | |||
| BC020043.1 | - | 2 | 269 | 4013548 | ||

Yeats4
Genomic Map