siRNA Details
| Gene Symbol: Etaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026576.3 | NP_080852.2 | 3, 4 | 524 | ||
| GenBank mRNA | AK019965.1 | - | 3, 4 | 525 | ||
| AK034138.1 | - | 3, 4 | 517 | |||
| BC028916.1 | - | 3, 4 | 519 | 3676092 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Etaa1| Gene Symbol: Etaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026576.3 | NP_080852.2 | 3, 4 | 524 | ||
| GenBank mRNA | AK019965.1 | - | 3, 4 | 525 | ||
| AK034138.1 | - | 3, 4 | 517 | |||
| BC028916.1 | - | 3, 4 | 519 | 3676092 | ||