siRNA Details
| Gene Symbol: Arl13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026577.3 | NP_080853.3 | 2, 3 | 422 | ||
| GenBank mRNA | AK016794.1 | - | 2, 3 | 368 | ||
| AK020859.1 | - | 2, 3 | 363 | |||
| AK021270.1 | - | 2, 3 | 423 | |||
| AK051159.1 | - | 2, 3 | 380 | |||
| BC082574.1 | - | 2, 3 | 422 | 30550373 | ||

Arl13b
Genomic Map