siRNA Details
| Gene Symbol: Cgnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB186125.1 | - | 19, 20 | 4310 | ||
| AK018850.1 | - | 2, 3 | 414 | |||
| AK019857.1 | - | 2, 3 | 401 | |||
| AK173252.1 | - | 16, 17 | 2638 | |||
| BC031499.1 | - | 2, 3 | 250 | |||
| BC039211.1 | - | 10, 11 | 1356 | |||
| BC139006.1 | - | 17, 18 | 3782 | 8862028 | ||
| BC158061.1 | - | 17, 18 | 3785 | 9007361 | ||

Cgnl1
Genomic Map