siRNA Details
| Gene Symbol: Hormad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289532.1 | NP_001276461.1 | 3 | 220 | ||
| NM_001289534.1 | NP_001276463.1 | 3 | 220 | |||
| NM_001289537.1 | NP_001276466.1 | 3 | 220 | |||
| NM_026489.3 | NP_080765.1 | 3 | 211 | |||
| GenBank mRNA | AK014945.1 | - | 3 | 219 | ||
| AK016324.1 | - | 3 | 219 | |||
| AK016743.1 | - | 3 | 214 | |||
| AK163995.1 | - | 3 | 219 | |||
| AY626343.1 | - | 3 | 219 | |||
| AY634284.1 | - | 3 | 219 | |||
| BC051129.1 | - | 3 | 219 | 6706580 | ||

Hormad1
Genomic Map