siRNA Details
| Gene Symbol: Apopt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163388.1 | NP_001156860.1 | 4 | 467 | ||
| NM_026511.2 | NP_080787.1 | 4 | 467 | |||
| GenBank mRNA | AK002873.1 | - | 4 | 469 | ||
| AK005403.1 | - | 4 | 455 | |||
| AK010493.1 | - | 4 | 457 | |||
| AK012648.1 | - | 4 | 465 | |||
| AK029408.1 | - | 4 | 443 | |||
| BC071262.1 | - | 4 | 467 | 6509633 | ||

Apopt1
Genomic Map