siRNA Details
| Gene Symbol: Fam173b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026546.3 | NP_080822.1 | 3, 4 | 458 | ||
| GenBank mRNA | AK020868.1 | - | 3, 4 | 458 | ||
| AK160923.1 | - | 585 | ||||
| BC030179.1 | - | 2, 3 | 449 | 5039149 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam173b| Gene Symbol: Fam173b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026546.3 | NP_080822.1 | 3, 4 | 458 | ||
| GenBank mRNA | AK020868.1 | - | 3, 4 | 458 | ||
| AK160923.1 | - | 585 | ||||
| BC030179.1 | - | 2, 3 | 449 | 5039149 | ||