siRNA Details
| Gene Symbol: Cntd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026562.2 | NP_080838.1 | 3 | 418 | ||
| GenBank mRNA | AK006754.1 | - | 3 | 204 | ||
| AK007245.1 | - | 3 | 419 | |||
| AK049116.1 | - | 3 | 526 | |||
| BC006866.1 | - | 3 | 477 | 3595113 | ||

Cntd1
Genomic Map