siRNA Details
| Gene Symbol: Atg101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026566.2 | NP_080842.1 | 2 | 478 | ||
| GenBank mRNA | AK007505.1 | - | 2 | 479 | ||
| AK150298.1 | - | 2 | 479 | |||
| AK151381.1 | - | 2 | 479 | |||
| AK160336.1 | - | 2 | 509 | |||
| AK167951.1 | - | 2 | 479 | |||
| BC016463.1 | - | 2 | 458 | 4166443 | ||

Atg101
Genomic Map