siRNA Details
| Gene Symbol: Fam135a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026604.4 | NP_080880.4 | 14 | 1893 | ||
| GenBank mRNA | AK034175.1 | - | 1 | 259 | ||
| AK044359.1 | - | 1 | 57 | |||
| AK053786.1 | - | 14 | 1812 | |||
| BC070446.1 | - | 13 | 1575 | 30362784 | ||

Fam135a
Genomic Map