siRNA Details
| Gene Symbol: Ccny | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026484.3 | NP_080760.2 | 10 | 1795 | ||
| GenBank mRNA | AK017493.1 | - | 10 | 1627 | ||
| AK028696.1 | - | 10 | 1796 | |||
| AK076556.1 | - | 10 | 1325 | |||
| BC023321.1 | - | 11 | 1713 | 3995040 | ||
| BC035524.1 | - | 9 | 1550 | 4974858 | ||

Ccny
Genomic Map