siRNA Details
| Gene Symbol: Coq5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026504.2 | NP_080780.1 | 7 | 1920 | ||
| GenBank mRNA | AK007496.1 | - | 7 | 1921 | ||
| AK045993.1 | - | 7 | 1928 | |||
| AK050334.1 | - | 7 | 1927 | |||
| BC052345.1 | - | 7 | 1927 | 6585618 | ||

Coq5
Genomic Map