siRNA Details
| Gene Symbol: Apopt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026511.2 | NP_080787.1 | 5 | 756 | ||
| GenBank mRNA | AK002873.1 | - | 5 | 758 | ||
| AK005403.1 | - | 5 | 744 | |||
| AK010493.1 | - | 5 | 746 | |||
| AK012648.1 | - | 5 | 754 | |||
| AK029408.1 | - | 5 | 732 | |||
| BC071262.1 | - | 5 | 756 | 6509633 | ||
| BY392793.1 | - | 1 | 79 | |||

Apopt1
Genomic Map