siRNA Details
| Gene Symbol: Mpnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026530.5 | NP_080806.4 | 11 | 1256 | ||
| GenBank mRNA | AK012128.1 | - | 10 | 1211 | ||
| AK146973.1 | - | 11 | 1241 | |||
| AK160356.1 | - | 11 | 1225 | |||
| AK172381.1 | - | 10 | 999 | |||
| AK216749.1 | - | 5 | 386 | |||
| BC017625.1 | - | 10 | 1205 | |||

Mpnd
Genomic Map