siRNA Details
| Gene Symbol: Psmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020132.1 | - | 7 | 1227 | ||
| AK159926.1 | - | 7 | 1183 | |||
| AK168886.1 | - | 6 | 1839 | |||
| BC004075.1 | - | 7 | 1186 | 3591796 | ||
| BC005717.1 | - | 7 | 1076 | 2609781 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmd8| Gene Symbol: Psmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020132.1 | - | 7 | 1227 | ||
| AK159926.1 | - | 7 | 1183 | |||
| AK168886.1 | - | 6 | 1839 | |||
| BC004075.1 | - | 7 | 1186 | 3591796 | ||
| BC005717.1 | - | 7 | 1076 | 2609781 | ||