siRNA Details
| Gene Symbol: Fam173b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026546.3 | NP_080822.1 | 5 | 1026 | ||
| GenBank mRNA | AK003724.2 | - | 2 | 555 | ||
| AK020868.1 | - | 5 | 1026 | |||
| AK044518.1 | - | 553 | ||||
| AK160923.1 | - | 1153 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam173b| Gene Symbol: Fam173b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026546.3 | NP_080822.1 | 5 | 1026 | ||
| GenBank mRNA | AK003724.2 | - | 2 | 555 | ||
| AK020868.1 | - | 5 | 1026 | |||
| AK044518.1 | - | 553 | ||||
| AK160923.1 | - | 1153 | ||||