siRNA Details
| Gene Symbol: Cntd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026562.2 | NP_080838.1 | 7 | 1177 | ||
| GenBank mRNA | AK007245.1 | - | 7 | 1178 | ||
| AK049116.1 | - | 7 | 1285 | |||
| BC006866.1 | - | 7 | 1236 | 3595113 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntd1| Gene Symbol: Cntd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026562.2 | NP_080838.1 | 7 | 1177 | ||
| GenBank mRNA | AK007245.1 | - | 7 | 1178 | ||
| AK049116.1 | - | 7 | 1285 | |||
| BC006866.1 | - | 7 | 1236 | 3595113 | ||