siRNA Details
| Gene Symbol: Wls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026582.4 | NP_080858.3 | 9 | 1461 | ||
| GenBank mRNA | AK019886.1 | - | 10 | 1500 | ||
| AK029053.1 | - | 9 | 1420 | |||
| AK029104.1 | - | 9 | 1418 | |||
| AK044810.1 | - | 9 | 1420 | |||
| AK049842.1 | - | 9 | 1418 | |||
| AK077407.1 | - | 9 | 1419 | |||
| AK128912.1 | - | 9 | 1417 | |||
| AK159207.1 | - | 9 | 1418 | |||
| BC018381.1 | - | 8 | 1045 | 4237619 | ||
| BC075615.1 | - | 9 | 1461 | 5703610 | ||

Wls
Genomic Map