siRNA Details
Gene Symbol: B230118H07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026592.3 | NP_080868.1 | 7 | 992 | ||
GenBank mRNA | AK051679.1 | - | 7 | 993 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B230118H07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026592.3 | NP_080868.1 | 7 | 992 | ||
GenBank mRNA | AK051679.1 | - | 7 | 993 |