siRNA Details
| Gene Symbol: D730048I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026593.3 | NP_080869.1 | 3 | 997 | ||
| GenBank mRNA | AK020302.1 | - | 3 | 969 | ||
| AK020310.1 | - | 3 | 950 | |||
| AK021351.1 | - | 3 | 969 | |||
| AK033813.1 | - | 3 | 998 | |||
| AK136636.1 | - | 3 | 968 | |||

D730048I06Rik
Genomic Map