siRNA Details
| Gene Symbol: Fam135a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026604.4 | NP_080880.4 | 14 | 2825 | ||
| GenBank mRNA | AK012197.1 | - | 1 | 310 | ||
| AK019549.1 | - | 1 | 443 | |||
| AK034175.1 | - | 1 | 1191 | |||
| AK044359.1 | - | 1 | 989 | |||
| BC031160.1 | - | 1 | 847 | |||
| BC070446.1 | - | 13 | 2507 | 30362784 | ||
| BC083093.1 | - | 1 | 304 | |||

Fam135a
Genomic Map