siRNA Details
| Gene Symbol: Leprotl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026609.2 | NP_080885.1 | 3 | 248 | ||
| GenBank mRNA | AK003329.1 | - | 3 | 259 | ||
| AK013870.1 | - | 3 | 318 | |||
| AK075740.1 | - | 3 | 303 | |||
| AK150289.1 | - | 3 | 307 | |||
| AK153277.1 | - | 3 | 293 | |||
| AK154712.1 | - | 3 | 313 | |||
| AK159401.1 | - | 3 | 236 | |||
| AK203783.1 | - | 2 | 206 | |||
| BC004677.1 | - | 3 | 248 | 3500223 | ||

Leprotl1
Genomic Map