siRNA Details
| Gene Symbol: Urm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026615.4 | NP_080891.1 | 2, 3 | 116 | ||
| GenBank mRNA | AK012124.1 | - | 2, 3 | 122 | ||
| AK019352.1 | - | 2, 3 | 122 | |||
| AK049227.1 | - | 2, 3 | 117 | |||
| AK049849.1 | - | 2, 3 | 117 | |||
| AK181129.1 | - | 100 | ||||
| AK210360.1 | - | 2, 3 | 100 | |||
| BC026994.1 | - | 2, 3 | 108 | 5368627 | ||

Urm1
Genomic Map