siRNA Details
Gene Symbol: Fam195a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026633.3 | NP_080909.1 | 4 | 391 | ||
GenBank mRNA | AK003564.1 | - | 4 | 393 | ||
AK020614.1 | - | 4 | 390 | |||
BC024332.1 | - | 4 | 383 | 5011430 |
